Would you like to log in again?
Essentra Wipes C100, FG Clean Wipes
Supplier: FG Cleanwipes
7C10077L00 7C10066P00
76239-594CS 502.66 USD InStock
76239-594 76238-330
Essentra Wipes C100, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FG clean wipes is a pioneer in the manufacturing of wipes for cleanrooms.
  • Significantly lower in particles than other nonwoven wiper fabrics
  • Pure and clean and residue free
  • Thicker, bulkier, three-dimensional structure absorbs and holds more liquids
  • Ideal for picking up aqueous spills
  • Very low extractables and ion levels
  • Textured structure increases particle removal
Wipes are hydroentangled and made of 51% cellulose and 49% polyester.
Suitable for ISO 5, 6, 7 and 8 environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty