Would you like to log in again?
Pharm-A-Line™ I Peristaltic-Pump Tubing, Thermoplastic Biologic (TBL)
Supplier: Thermoplastic Biologic (TBL)
PHI-031X093-50
76252-080EA 220.3 USD InStock
76252-080
Pharm-A-Line™ I Peristaltic-Pump Tubing, Thermoplastic Biologic (TBL)
Tubing Transfer Tubing
The Tubing is manufactured from virgin, medical grade TPE and homogeneously extruded to precise dimensional tolerances, specifically developed to provide superior performance in peristaltic pump applications.
  • Multiple autoclave cycles
  • Lot and batch tractable
  • Low particulate and USP Class VI
It provides an excellent alternative to silicone tubing when resistance to chemicals (such as acetone or methylene chloride) is a concern. Ideal for pharmaceutical, biological, and food and beverage applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty