Would you like to log in again?
3M™ Double Coated Urethane Foam Tape
Supplier: 3M
3M™
7000048483 7000048486 7000049238 7000048482 7000048484
76245-010CS 1300.15 USD InStock
76245-010 76243-258 76242-344 76251-914 76242-350
3M™ Double Coated Urethane Foam Tape
Tapes
These tapes are thick double coated, open-cell urethane foam tape with a paper liner.
  • High shear strength
  • Bonds to irregular surfaces
  • Smooth, clean appearance
  • Even stress distribution
  • Quick stick
These are conformable foam tapes that features high ultimate bond strength, very good high temperature and solvent resistance and very high shear holding power. A natural choice for indoor mounting, joining and holding applications. It bonds and fills gaps, including on irregular surfaces, and provides excellent handling convenience and cushioning.
Applications include general Industrial, bonding, attaching and mounting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty