Would you like to log in again?
PREpleat LPD HC Filter MERV 8, Flanders Filters, American Air Filter Company
Supplier: AMERICAN AIR FILTER COMPANY TE
80255.0108H61B
76240-212CS 144.93 USD InStock
76240-212
PREpleat LPD HC Filter MERV 8, Flanders Filters, American Air Filter Company
Filters Gas Filters
Low resistance MERV 8 media, providing higher DHC than competitive high capacity filters.
  • Ecologically friendly frame components made from recyclable materials
  • Expanded metal grid prevents media flutter while in operation
  • Filter media pack is sealed to eliminate air bypass
  • MERV 8 high capacity
Superior frame strength due to diagonal and horizontal support members, as well as wire-backed media.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty