Would you like to log in again?
PREpleat 40 Economy Filter MERV 7, Flanders Filters, American Air Filter Company
Supplier: AMERICAN AIR FILTER COMPANY TE
80055.0118C18C
76245-690CS 46.19 USD InStock
76245-690
PREpleat 40 Economy Filter MERV 7, Flanders Filters, American Air Filter Company
Filters Gas Filters
PrePleat 40 economy filters, that is more efficient and ecologically friendly. Made entirely from recyclable materials, this medium achieves MERV 7 efficiency with low resistance to airflow. It is also unaffected by high humidity and is hydrophobic (non-moisture absorbing).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty