Would you like to log in again?
Precision Electronic Test Probe Set, TESTEQUITY LLC TE
Supplier: TESTEQUITY LLC TE
6275
76268-200EA 114.19 USD InStock
76268-200
Precision Electronic Test Probe Set, TESTEQUITY LLC TE
Electrodes Other Electrodes
Precision Electronic Test Probe Set.
  • Replaceable 0.04" (1.01 mm) pogo-pin tips
  • Small outline for tight test situations
  • Comfort-grip textured surface
Fits all meters including Fluke, HP (Agilent), Keithley, Meterman and silicone insulated leads resist abrasion and heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty