Would you like to log in again?
Wire Marking Sleeves, PermaSleeve®, Brady
Supplier: Brady
PermaSleeve
32607 110412 112535 142096
76253-218EA 713.73 USD InStock
76253-218 76251-020 76251-024 76251-028
Wire Marking Sleeves, PermaSleeve®, Brady
Labels
These PermaSleeve® PS heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high quality identification.
  • Fade-resistant and smudge-proof for superior aesthetic appearance
  • With thermal transfer print technology
  • Cold resistant and flame-retardant with an operating temperature of –55 to 135 °C (–67 to 275 °F)
These heat shrinkable sleeves fit snugly around wires for maximum insulation protection and permanent identification.
Certifications: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202; RoHS Compliant; and UL Recognized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty