Would you like to log in again?
Self Laminating Labels, Brady
Supplier: Brady
111356 111328 111340 111357 111354 111351 111358 111355
76249-026PK 171.34 USD InStock
76249-026 76249-036 76249-024 76249-034 76249-022 76249-032 76249-030 76249-028
Self Laminating Labels, Brady
Labels
These labels features B-292 material which has good clarity outstanding water and oil resistance.
  • Vinyl labels feature clear trailer that wraps to protect printed text
  • Flexible, self-extinguishing material conforms to most surfaces
  • Operating temperature of –40 to 65.5 °C (–40 to 150 °F)
Ideal for use in wire and cable identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty