Would you like to log in again?
Wire, Cable Labels, Brady
Supplier: Brady
111751 112225 112063
76242-286PK 142.96 USD InStock
76242-286 76250-238 76242-284
Wire, Cable Labels, Brady
Labels
These B-500 repositionable vinyl cloth wire markers are perfect for general wire marking needs.
  • Card-mounted wire markers allow for quick, on-the-spot wire marking
  • Excellent flexibility for wrapping around curved surfaces
  • Withstands moderate heat, oil, water and dirt
The vinyl cloth holds tightly to surfaces but allows item to be repositioned with ease if necessary.
Abrasion, corrosion, humidity, oil, UV, water and weather-resistant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty