Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CPU Holder, Arlink

Supplier: Arlink

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

ARL-8440
76267-964EA 348.05 USD InStock
76267-964
CPU Holder, Arlink
Computer and Equipment Stands and Mounts
Adjustable pullout CPU holder has storable slide travel of 19".

  • 360° rotation
  • Accommodates CPU width range of 2 to 812" and height range of 11 to 22"

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty