Would you like to log in again?
Lista Plastic Laminate Worksurface Assemblies for Arlink® 8000 Workstations, Arlink
Supplier: Arlink
Lista™ Xpress
ARL-SA101 ARL-8730 ARL-8265
76240-252EA 0.73 USD InStock
76240-252 76240-250 76268-294
Lista Plastic Laminate Worksurface Assemblies for Arlink® 8000 Workstations, Arlink
Furniture Tables and Workbenches
Worksurface is constructed of 1" thick medium density particle board covered on top and all sides with non-glare matte finish plastic laminate.
  • Front edge is contoured for comfort and chip resistance
  • Threaded steel inserts facilitate easy assembly and reconfiguration
The under side of the worksurface is sealed with a plastic tension sheet to prevent moisture absorption, warping and shedding.
Support leg attaches to front support beam of worksurface to increase load capacity or dampen vibration.
Delivery information: Assembly includes two worksurface support brackets and two worksurface support beams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty