Would you like to log in again?
Self-Laminating Vinyl Labels for Thermal Transfer Printers, Brady
Supplier: Brady
106452 30550 62352 18365 18434 18341 18346 131571
76248-974EA 208.04 USD InStock
76248-974 76247-504 76244-724 76242-078 76245-016 76242-074 76247-742 76242-076
Self-Laminating Vinyl Labels for Thermal Transfer Printers, Brady
Labels
Self-laminating vinyl material provides a protective layer over text for excellent abrasion, dirt, oils, solvents, water and excess handling resistance.
  • Service temperature of −40 to 158 °F (−40 to 70 °C)
  • Ideal for use in cable and wire labeling, voice-data identification, and panel marking
  • Printable white zone with self-lamination prints on a white background with translucent over laminating area to protect text
Permanent acrylic tape adheres well to surfaces that are smooth, rough or powder coated, and to irregular surfaces or surfaces with low surface energy.
Resistant to chemical, humidity and weather.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty