Would you like to log in again?
Micro NMR Sample Tubes, New Era™
Supplier: New Era
NEH5/3 NEH5/2.5
100165-950EA 49.72 USD InStock
100165-950 100165-948
Micro NMR Sample Tubes, New Era™
Tubes NMR Tubes
Micro tubes are designed for use with reduced sample volumes in cryo/cold probes as well as micro and all standard 5 mm probes.
  • Uniform sample column
  • Tube configurations for most applications
  • Attain spin rate and auto-shim quickly
  • Collect the best data in the shortest time
  • More efficient use of spectrometer time
The body also accommodates the Teflon adapters used to secure a central capillary to further reduce the sample volume. Narrow sample column eliminates any susceptibility issue. Coaxial alignment is maintained to give the most stable sample column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty