Would you like to log in again?
Sleeve Cap for Tube Ends, SC Series, Caplugs
Supplier: Caplugs
SC-1/2-8
76244-504PK 448.17 USD InStock
76244-504
Sleeve Cap for Tube Ends, SC Series, Caplugs
Closures Safety Caps and Safety Waste Caps
SC Series sleeve caps are designed to fit outside diameters of tubing with a snug friction grip.
  • Ideal for capping and protecting both threaded and non-threaded tubing and rod ends
  • Made of low-density polyethylene
It also make excellent masks for painting, plating or grit-blasting operations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty