Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Density Poly Liner with Star Seal, Associated Bag

Supplier: ASSOCIATED BAG INC.
52-200
76247-920EA 103.87 USD InStock
76247-920
High Density Poly Liner with Star Seal, Associated Bag
Bags Waste Disposal Bags
High density poly liner with star seal on a coreless roll (8 µm).

  • Dispenser box prevents liners from unrolling
  • Liners stay clean and organized
  • Use with 7½ to 10 gallon cans, circumference of 48"

Lightweight material is twice as strong as conventional poly. Strong, star-designed bottom seal is gathered and sealed tight in multi-layers to eliminate leaks.

Delivery information: Ties included.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty