Would you like to log in again?
Copper Foil Shielding Tape, 3M
Supplier: 3M
7010398771
76254-074CS 1629.83 USD InStock
76254-074
Copper Foil Shielding Tape, 3M
Tapes
Cap has a 1.4 mil copper foil backing with conductive acrylic pressure-sensitive adhesive on both sides and a liner.
  • Excellent conductive gasket
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty