Would you like to log in again?
Diamond Wheel Cutter, Dremel
Supplier: Dremel
545
76246-918EA 82.38 USD InStock
76246-918
Diamond Wheel Cutter, Dremel
Cutters
Diamond wheel is completely coated with fine diamond particles for working with hard materials. Designed to be long lasting and not lose its shape or break, it provides smooth, fine cuts.
  • Not recommended for metal
Ideal for cutting, sawing, carving, sharpening and filing hard materials-including marble concrete, brick, porcelain, ceramics, epoxy and hard woods.
Caution: Do not run in excess of 20000 rpm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty