Would you like to log in again?
Scorpion2 Power Supply and Cable Kit, Simcona Electronics
Supplier: Simcona
5051328
76247-600EA 352.35 USD InStock
76247-600
Scorpion2 Power Supply and Cable Kit, Simcona Electronics
Power Supplies
Power supply kit that is ideal for ionizing bars. The power supply and cable kit are designed for a wide range of applications.
  • Air assist version
  • Peak reduction technology
  • Daisy chain feature with master/slave
For use with Scorpion3 ionizing bar.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty