Would you like to log in again?
Dri-Shield® Moisture Barrier Bag, 2700 Series, SCS, Desco Industries
Supplier: Desco Industries
D27810 D2768
76245-396PK 100.2 USD InStock
76245-396 76241-332
Dri-Shield® Moisture Barrier Bag, 2700 Series, SCS, Desco Industries
Bags Sample Bags
These aluminized moisture barrier bags are designed to provide a static safe environment for sensitive electronic devices.
  • Stronger polyester layer prevents sharp trays from puncturing the bag
  • Heat sealable
  • Suitable for vacuum packaging
  • Free of amines, silicones and heavy metals
  • Provides superior physical protection for contents
Bags are printed with ESD and moisture warning symbols and a lot code for traceability. Manufactured from a laminate of multiple layers of aluminized polyester and polyethylene. Polyester provides puncture resistance. Metal layers are intended to provide shielding of Electrostatic Discharge (ESD) and to help minimize the penetration of electric field.
Designed for dry packing of moisture sensitive devices and to protect contents from damage due to ESD events or EMI.
Certifications: Meets ANSI/ESD S20.20, ANSI/ESD S541, EIA 541, JESD625-A, and EIA 583.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty