Would you like to log in again?
Valve Tags, Blank, Brady
Supplier: Brady
44403
76244-922PK 188.94 USD InStock
76244-922
Valve Tags, Blank, Brady
Labels
Blank stainless steel tags are pre-punched with a ³/₆" hole (0.188" Int.Ø).
  • Tags are ideal for harsh environments or where stainless steel material is specified
  • Service temperature range of –40 to 1500° F
  • Average outdoor durability of 5 to 7 years
B-919 Stainless steel tags are used for general purpose valve identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty