Would you like to log in again?
Chicago Latrobe® NAS-Type Cobalt Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Supplier: Cutting Tool Supply
46604 54078 46683 46609 46691
76249-088EA 12.82 USD InStock
76249-088 76250-098 76243-932 76242-876 76243-260
Chicago Latrobe® NAS-Type Cobalt Heavy-Duty Jobber Length Drills, Cutting Tool Supply
Tool Sets
Heavy-duty construction for drilling in tough, high-tensile, and work-hardening materials under extreme operating conditions.
  • 135° P3 split point is self-centering for reduced thrust and easier penetration
  • Manufactured from premium cobalt high-speed steel
  • Increasing hot hardness and extending drill wear life
Straw finish adds lubricity and easily identifies cobalt drills and titanium nitride (TiN) coating adds lubricity and hardness, enhancing chip flow, finish hole quality, and drill life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty