Would you like to log in again?
VitroCol® Human Collagen
Supplier: Advanced BioMatrix
VitroCol®
5008-15MG
103700-652EA 392.31 USD InStock
103700-652
VitroCol® Human Collagen
Proteins and Peptides
VitroCol® collagen is the first widely available, naturally produced purified human collagen for research purposes. VitroCol® sets the standard for purity (>99% collagen content), functionality and represents the only native-like human collagen offered.
VitroCol® collagen is naturally secreted from human neo-natal fibroblast cells. The human fibroblasts are cultured in optimal conditions allowing the fibroblasts to naturally and efficiently secret extracellular matrix. The extracellular matrix is then processed and purified to yield the naturally produced human collagen.
VitroCol® is approximately 97% Type I human collagen with the remainder being comprised of Type III collagen. It contains high monomer content as measured by gel permeation chromatography.
This product is supplied as a lyophilized powder with 15 mg of human collagen. When reconstituted with 5 ml of sterile 0.01 N HCl, a concentration of approximately 3 mg/ml is achieved.
VitroCol® is especially ideal for human cell culture systems when coating of surfaces and providing preparations of thin layers of culturing cells. VitroCol®, lyophilized form is not recommend for the formation of a solid gel. VitroCol® human collagen is provided in user-friendly packaging for use and storage. VitroCol® is supplied as a sterile, lyophilized powder.
  • Great for 2D coatings
  • Highly purified, native ECM
  • Known for purity, consistency and quality
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty