Would you like to log in again?
PureCol® Bovine Collagen
Supplier: Advanced BioMatrix
PureCol®
5015-20ML 5005-100ML
103700-632EA 333.46 USD InStock
103700-632 103700-638
PureCol® Bovine Collagen
Proteins and Peptides
PureCol® -S collagen is provided as a standard for use in assays where ultra-pure collagen is required. PureCol®-S collagen standard is approximately 97% Type I collagen with the remainder being comprised of Type III collagen. It contains a high monomer content as measured by gel permeation chromatography. This product is supplied at approximately a 3 mg/ml concentration. The concentration is confirmed by Biuret protein determination assay. The concentration for each specific lot is provided on a Certificate of Analysis that is available with the purchase of each product. PureCol®-S is soluble atelo-collagen in 0.01 N HCI, therefore, the pH is approximately 2.0.
PureCol® -S is ideal for using as a collagen standard in controlled testing and assay procedures. PureCol®-S is sterile filtered and is supplied as a ready to use solution.
  • Great for 3D Hydrogels or 2D Coatings
  • Highly purified collagen
  • Over 15 QC tests to ensure consistency and quality
  • Recognized worldwide collagen standard
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty