Would you like to log in again?
Peptite-2000® Peptide-2000
Supplier: Advanced BioMatrix
5020-5MG
103701-186EA 398.85 USD InStock
103701-186
Peptite-2000® Peptide-2000
Proteins and Peptides
PEPTITE-2000® is a synthetic peptide containing the RGD cell attachment sequence found in fibronectin, vitronectin and many other matrix and serum proteins. This RGD peptide has been specifically engineered to spontaneously adhere to a variety of surfaces such as polystyrene (either tissue culture treated or non-treated), hollow fibers made of cellulose, glass, polycarbonate membranes and many other materials. This binding is mediated via a hydroscopic C-terminal sequence. The RGD motif is present at the N-terminal end of the peptide, allowing for optimal cell attachment via integrin receptors.
PEPTITE-2000® can be substituted as the primary cell attachment component in a chemically defined animal component-free culture system. PEPTITE-2000® is supplied in sterile 5 mg quantities.
  • Great cell binding peptide
  • Irradiation Sterilized
  • Animal-free
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty