Would you like to log in again?
Static Dissipative Polyimide Labels for Thermal Transfer Printers, UltraTemp® B-717, Brady
Supplier: Brady
UltraTemp UltraTemp®
134243 134237 134241
76245-836EA 405.45 USD InStock
76245-836 76245-952 76248-344
Static Dissipative Polyimide Labels for Thermal Transfer Printers, UltraTemp® B-717, Brady
Labels
Static dissipative polyimide withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labeling.
  • Service temperature of –94 to 212 °F (–70 to 100 °C)
High temperature, glossy white polyimide material features a static dissipative adhesive and static dissipative release liner.
Certifications: Surface resistivity values in the recommended range for Dissipative ESD packaging materials as defined by ANSI/ESD S541-2008. RoHS compliant to 2011/65/EU; UL Approved; Meets requirements of MIL-STD-202G, Method 215K.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty