Would you like to log in again?
Morse® 2-Flute Miniature Long Length Double End Mills, Cutting Tool Supply
Supplier: Cutting Tool Supply
43253-MO
76249-142EA 94.78 USD InStock
76249-142
Morse® 2-Flute Miniature Long Length Double End Mills, Cutting Tool Supply
Tool Sets
These miniature center cutting shank end mills are designed for small diameter milling of slots, keyways and pockets.
  • Shorter length provides maximum rigidity
Center cutting end allows for plunge cutting like a drill into solid material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty