Would you like to log in again?
Round Vinyl Caps, VC Series, Caplugs
Supplier: Caplugs
VC-1062-16 VC-062-8 VC-093-8 VC-062-10 VC-080-8-RED VC-234-8 VC-080-8-BLK VC-937-16
76247-648PK 497.46 USD InStock
76247-648 76245-550 76248-778 76249-372 76248-172 76248-276 76248-770 76247-212
Round Vinyl Caps, VC Series, Caplugs
Fittings and Connectors
VC series caps are extremely flexible and durable protective closures which are ideal for use as thread protectors, dust and moisture seals, pipe and stud end color coding, and for masking during painting or plating operations.
  • Wide range of sizes
  • Easy to use
  • Vast color range available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty