Would you like to log in again?
PureFlo® SKL Series PP Media Capsule, Zenpure Americas
Supplier: ZENPURE AMERICAS C/O ST GOBAIN TE
SKLP010STCTC
76247-656EA 196.14 USD InStock
76247-656
PureFlo® SKL Series PP Media Capsule, Zenpure Americas
Filters Capsule Filters
PureFlo® Polypropylene Media filter capsules are highly retentive, graded porosity, pleated polypropylene media filters that have been specially designed for clarification and prefiltration applications.
  • High flow rates and low pressure drops
  • Excellent thermal and chemical compatibility
  • Superior filter lifetime and process throughputs
  • Consistent and reproducible particulate removal
  • Rapid installation and reduced downtime
  • Reduced chemical handling and hazardous waste disposal
The graded porosity design removes particles in sequence by size – larger particles by the more open outer layers and smaller particles by the tighter inner layers. The outer layers act as a pre-filter while the inner layers provide the absolute rating at the specified pore size. This efficiently spreads the contaminants throughout the media matrix resulting in superior contaminant holding capacity, lifetime, and pressure drop as compared to other media cartridges.
No adhesives, binders, or surfactants are used in the manufacturing process The non-woven media does not allow for migration into the process fluid, thereby reducing the potential for extractables and downtime. Furthermore, the allpolypropylene construction provides excellent thermal and chemical compatibility with low and high pH chemicals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty