Would you like to log in again?
Scotch® Filament Tape 898, 3M™
Supplier: 3M
7000001229 7000028930
76242-416CS 2381.98 USD InStock
76242-416 76243-650
Scotch® Filament Tape 898, 3M™
Tapes
Scotch® Filament tape is a high-performance transparent packaging tape reinforced along its length with continuous glass-yarn filaments, which give the tape very high tensile strength.
  • Transparent polypropylene backing provides excellent abrasion, moisture and scuff resistance
  • Allows printing and illustrations to be seen through the tape
  • High shear and great initial adhesion
  • Excellent aging
  • Good protection of filaments and adhesive to provide longer package life
  • Assures maximum package performance for limited outdoor use
Adhesive is an aggressive packaging adhesive specifically formulated to provide balanced performance having both high adhesion and long term holding power.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty