Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcohol reagent ≥100% ACS

Supplier: Avantik
Synonyms: Reagent alcohol

RS4029
76270-152CS 107.65 USD InStock
76270-152
Alcohol reagent ≥100% ACS
Alcohol
A.C.S. grade. Meets and
exceeds histological grade.

Avantik Alcohol is made from A.C.S. Grade Reagent Alcohol. The lower alcohol content varieties are made with A.C.S. Grade 100% Alcohol. The highest quality for all histology processing and staining procedures.


MDL Number: MFCD00003568
CAS Number: 64-17-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty