Would you like to log in again?
Weller® Precision 115 mm Tweezer with Pointed Tips, Apex Tool Group
Supplier: Apex
Weller®
5SA
76238-614EA 128.88 USD InStock
76238-614
Weller® Precision 115 mm Tweezer with Pointed Tips, Apex Tool Group
Forceps
Precision tweezer is designed for fine wires.
  • For precision work under a microscope
  • Pointed tips, suitable for very fine wires
  • Relieved shape facilitates excellent access to the most confined spaces
The special stainless steel is non-magnetic. The tweezers are non-rusting, acid-proof and heat resistant up to 300 °C (512 °F). Tweezers made from special stainless steel have the identification letter "SA" in their order numbers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty