Would you like to log in again?
Aluminum Handle Scratch Brushes, Gordon Brush
Supplier: Gordon Brush
33HHA 22HHA
76249-310PK 328.2 USD InStock
76249-310 76243-290
Aluminum Handle Scratch Brushes, Gordon Brush
Brushes
Brushes with anti-static horsehair bristle and a hand-laced aluminum handle.
  • Horsehair fill material is hand-laced with stainless steel wire into its handle for maximum fiber retention
  • With two or three rows of bristles
  • Anti-static
Ideal for detail cleaning on items such as printed circuit boards and SMT assemblies, or for use as fluid applicators. Suitable for use in an ESD environment provided the brush remains saturated in liquid. It is recommended that the brush be tested on the required application to confirm ESD compatibilty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty