Would you like to log in again?
PHS-1KBIR Preheater Set, JBC
Supplier: JBC
PHSE-1KB
76248-794EA 4477.27 USD InStock
76248-794
PHS-1KBIR Preheater Set, JBC
Soldering Supplies Desoldering Tools
Complete system for preheating multilayer PCBs and ideal for repetitive soldering jobs.
  • Perfectly adapted to any type of PCBs
  • Three adjustable height positions
The infrared preheater PHS is used as a secondary heat source to reduce the amount of energy supplied by the main soldering tool, reducing thermal shock to components and PCBs.
Heating area is measured as 6.5×13.5 cm for one zone and 13×13.5 cm for two zones.
Delivery information: Consists of close loop thermocouple controlled infrared preheater unit and with a PCB support.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty