Would you like to log in again?
Cup Style Heel Grounder, SCS, Desco Industries
Supplier: Desco Industries
HGC1M
76238-796EA 21.59 USD InStock
76238-796
Cup Style Heel Grounder, SCS, Desco Industries
Grounders Heel and Toe Grounders
The re-positionable configuration of the Heel Grounder extends the life of the product. An elastic loop provides a secure and comfortable fit.
  • Wide sole provides additional surface area for a better path to ground
  • Tear resistant rubber with inner scrim; reinforced material prevents tearing and increases the service life
  • Elastic hook and loop strap
  • Size adjustable material expands to fit most shoe sizes comfortably and flex during walking
  • Non-marking interior layer will not mar operator’s shoes
  • 1 megohm series resistor protects operator from accidental contact with equipment line voltage
  • Made in the United States of America
This quality Heel Grounder features a durable three-layer sole. A 24" conductive ribbon grounding tab provides additional conductive material for reliable connection with operator’s sweat-layer. Lot coded for QC traceability.
Certifications: Rg: <1.2×106 ohms per ESD SP9.2 7 ESD TR53. UL listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty