Would you like to log in again?
KEMRESIN® Venturi Work Tops, Kewaunee®
Supplier: Kewaunee
KEMRESIN®
VBTR012460-A VBTR012472-AC VBTR012460-AC VBTR012448-A VBTR012496-AC VBTR012472-A VBTR012448-AC VBTR012496-A
KVBTR012472-ACEA 2217.95 USD InStock
KVBTR012472-AC KVBTR012460-AC KVBTR012496-AC KVBTR012448-AC KVBTR012496-A KVBTR012472-A KVBTR012460-A KVBTR012448-A
KEMRESIN® Venturi Work Tops, Kewaunee®
Fume Hoods and Enclosures Chemical Fume Hoods
Venturi fume hood worktop can be used wherever optimum physical and chemical resisting properties are required.
  • Ultra-smooth non-porous surface
  • Composed of 1" and ³/₄" thick molded epoxy resin
  • Resistant to all alkalis, acids and solvents
  • Highly resistant to heat and physical abuse
It also contains features such as left rear cupsink, horizontal flush.
Certifications: Meet SEFA 3 chemical resistance performance standards.
Ordering information: Available in the colors: black, putty, gray, slate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty