Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Pyrrolidone ≥99%

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Pyrrolidin-2-one

P2121-200LTBL P2121-500MLGL P2121-100ML
76014-820EA 7114 USD InStock
76014-820 75873-212 75873-214
2-Pyrrolidone ≥99%
2-Pyrrolidone


Formula: C₄H₇NO
MW: 85.11 g/mol
Density: 1,120
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
Order Now
Specification Test Results
Assay Min. 99%
Color (APHA) Max. 100
Moisture 0.5%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty