Would you like to log in again?
SIRUS® Non-Sterile Poly-Reinforced Gowns, Medline Industries
Supplier: Medline
SPT-2208SMSCS
76246-194CS 1679.06 USD InStock
76246-194
SIRUS® Non-Sterile Poly-Reinforced Gowns, Medline Industries
Disposable Garments
Dependable protection from five fabric layers, three densely-packed meltblown layers sandwiched between two strong spunbound outer layers, for an excellent barrier against fluids and pathogens. Medline's Sirus® line offers reliability and comfort. AAMI Level 4 protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty