Would you like to log in again?
Syringe Caps, OK International
Supplier: OK International
730000CBPK
76248-730PK 104.68 USD InStock
76248-730
Syringe Caps, OK International
Closures Safety Caps and Safety Waste Caps
End cap design to seal inlet side of syringe barrel to ensure no contaminants come in contact with material during storage and shipment.
Packaging: Packaged in box of 1000 pieces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty