Would you like to log in again?
Filter Cartridge, VP Series, H2O Filter Warehouse
Supplier: H2O FILTER WAREHOUSE TE
VP-17-4011
76242-046CS 480.93 USD InStock
76242-046
Filter Cartridge, VP Series, H2O Filter Warehouse
Filters Filter Cartridges
This VP series cartridg provide ultra high purity filtration for a variety of applications. Constructed of non-pigmented, virgin materials the spin welded construction provides the longest life and highest quality.
  • Hose nipple end caps have tapered ends for easy connections
  • Straight nipple tube ends can be used with any ³/₈" compression or tubing fitting
Filter length can be easily modified for custom applications and clear tubes are available for color changing medias. Combination cartridges offer multiple stages of filtration in a single filter.
Applications include deionization, submicron filtration, organics, chlorine, oxygen and amine removal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty