Would you like to log in again?
Mantis Compact Stereo Microscope, Vision Engineering
Supplier: Vision Engineering
MCS-005 MCH-001
76243-834EA 2136.15 USD InStock
76243-834 76243-718
Mantis Compact Stereo Microscope, Vision Engineering
Microscopes
Mantis Compact is an ergonomic, low fatigue visual inspection microscope, excelling in the low magnification range for inspection tasks where bench magnifiers have traditionally been used.
  • 2x, 4x, 6x and 8x quick change magnification options
  • Superior ergonomics for fatigue-free viewing and increased quality / productivity
  • Superb hand-eye co-ordination for inspection and manipulation tasks
  • Long working distance and large field of view
  • True colour, LED illumination providing up to 10,000 hours of shadow-free viewing
Featuring Vision Engineering’s patented eyepiece-less technology, Mantis Compact is the low magnification stereo viewer of choice for a wide range of inspection, manipulation or rework tasks requiring magnification.
Patented optical technology not only provides superb 3D imaging, but also allows operators freedom of head movement.
Superior ergonomics leads to greater productivity, increased throughput, improved quality control as well as minimal fatigue. Users are also able to wear glasses, if required.
Mantis Compact is ideal for a wide range of tasks requiring 3D imaging with superb hand-to-eye co-ordination, including electronics inspection, dental re-work, dissection and preparation tasks, plus many other precision magnification tasks.
Ordering information: Head and stand need to be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty