Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc acetate dihydrate 99-102%, Puriss., meets analytical specification of DAB, E650, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Zinc diacetate dihydrateAcetic acid zinc salt dihydrateZinc (II) acetate dihydrate

25044-1KG
BJ25044-1KGEA 119.38 USD InStock
BJ25044-1KG
Zinc acetate dihydrate 99-102%, Puriss., meets analytical specification of DAB, E650, Fluka™
Zinc acetate dihydrate


Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 100 °C
Density: 1.750 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
ADR: 9,III
Order Now
Specification Test Results
Assay 99.0 - 102.0 %
pH (5 %, 25 °C) 6.0 - 8.0
Water insoluble matter Max. 0.005 %
in CH₃COOH insoluble matters Max. 0.005 %
Alkalis/earth alkalis Max. 0.2 %
Arsenic (As) Max. 3 ppm
Calcium (Ca) Max. 0.005 %
Cadmium (Cd) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.005 %
Lead (Pb) Max. 20 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 100 ppm
Organic volatile impurities (GC) complying
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty