Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Anti-V5-tag mAb-Magnetic Beads, MBL International

Supplier: MBL International
M167-11
103268-788EA 377.89 USD InStock
103268-788
Anti-V5-tag mAb-Magnetic Beads, MBL International
Beads Magnetic Beads
Monoclonal antibody conjugated to magnetic beads of 20 tests targeting V5-tag for IP.

  • Antibody type: Monoclonal
  • Formulation: 10 mg magnetic beads in 1 ml PBS/0.1% BSA/0.09% NaN₃
  • Host species: Mouse
  • Immunogen: Carrier protein conjugated synthetic peptide, GKPIPNPLLGLDST (V5-tag)
  • Isotype: IgG2a k
  • Storage temperature: 4 °C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty