Would you like to log in again?
Preservation Sealing Tapes, 3M
Supplier: 3M
7000048405
76253-830CS 2187.44 USD InStock
76253-830
Preservation Sealing Tapes, 3M
Tapes
Preservation sealing tape 481 seals and helps to protect various types of equipment to be stored by military and industrial manufacturers. Tapes helps to resist weathering, conforms to many irregular surfaces and has excellent stain resistance.
  • Easily conforms to a irregular surfaces
  • Excellent resistance to staining, common lubricants and strippable coatings
  • Excellent weatherability for preserving equipment and vehicles
The tapes removes cleanly after up to two years outdoor weathering.
Certifications: SAE-AMS-T-22085 Type II.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty