Would you like to log in again?
STOP Safety Glasses Required Beyond This Point Sign, Brady
Supplier: Brady
124543
76252-656EA 73.81 USD InStock
76252-656
STOP Safety Glasses Required Beyond This Point Sign, Brady
Signs
Plastic signs are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • High impact polystyrene with protected graphics
  • Good abrasion and chemical resistance
Recommended for both indoor and outdoor use. Service temperature of –40 to 175 °F (–40 to 80 °C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty