Would you like to log in again?
APSH Braid-Reinforced Silicone Hose, Newage Industries
Supplier: NewAge Industries
APSH-P-0250
76240-198EA 1249.07 USD InStock
76240-198
APSH Braid-Reinforced Silicone Hose, Newage Industries
Tubing Transfer Tubing
This low volatile grade, platinum-cured silicone hose is clean-room produced for critical pharmaceutical, biomedical, cosmetic, and food applications.
  • Not suitable for continuous steam applications
  • Hardness value of 65 Shore A
  • Extremely flexible
  • Imparts no taste or odor to critical streams
  • Sterilizable by autoclave, CIP, SIP, and gamma radiation processes
  • Documented lot traceable with identification on bags
Polyester yarn braiding inside the wall enhances pressure capabilities. This hose resists temperature extremes, compression set, chemical attack, ozone, radiation, moisture, and environmental exposure.
Certifications: APSH has undergone extensive physical, chemical, and biological testing and meets USP Class VI, FDA CFR 177.2600, ISO 10993, European
Pharmacopoeia 3.1.9, and 3-A standards. Listed by the National Sanitation Foundation for food equipment materials (NSF-51). APSH has a master file with the U.S. Food and Drug Administration (#27582).
Packaging: Supplied double bagged, other packaging options available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty