Would you like to log in again?
980 Ionized Air Gun, SCS Division of Desco
Supplier: Desco Industries
980
76238-102EA 3213.11 USD InStock
76238-102
980 Ionized Air Gun, SCS Division of Desco
Ionizers
This air gun neutralizes electrostatic charges to eliminate ElectroStatic Attraction (ESA), visual imperfections, and contamination by dislodging charged dust and debris.
  • Modular design with seven foot esd protective hose with trigger switch
  • Allows for quick change between compressed gas supply
  • Quick filter replacement feature minimizes down time
This allows operators freedom of motion and minimizes awkward or unsupported positions.
Certifications: NIST calibrated with certificate included. Meets ANSI/ESD S20.20 ionizer required limits, cULus, CE, RoHS2.
Accessories information: Replacement emitter tip sold separately. Use with 980 Ionized Air Gun and 980E Ionized Air Gun.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty