Would you like to log in again?
Electric Screwdrivers, HIOS CL, Mountz
Supplier: Mountz
144116 144121
76251-268EA 3456.97 USD InStock
76251-268 76251-266
Electric Screwdrivers, HIOS CL, Mountz
Screwdrivers
The CL-Series electric torque screwdriver is designed for heavy-duty production and delivers reliability as well as precision torque.
  • Low DC voltage for safe operation and long life
  • Precision automatic shut-off clutch once preset torque is achieved
  • Compact size with a grip designed for operator comfort
  • External torque adjustment scale
  • External brush replacement means no downtime for brush replacement
  • Torque levels can be easily adjusted by turning down the external adjustment ring
  • Connectable with the Scout screw counter
Accurate torque control provides precise and controlled fastening with automatic shut-off clutch. The full auto shut-off clutch on this electric screwdriver completely stops the bit from turning once the torque setting is reached. This feature protects the fastener from being damaged by the electric screwdriver's bit.
Common usages for this electric screwdriver include demanding assembly-fastening applications that require torque control, screw counting functionality and ESD protection.
Certifications: Drivers meet electrical safety standards in the U.S. and Europe (UL Listed and CE Certified).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty