Would you like to log in again?
3M™ Scotch-Weld™ Metal Primer
Supplier: 3M
7000000907
76245-582CS 1667.62 USD InStock
76245-582
3M™ Scotch-Weld™ Metal Primer
Adhesives
3M™ Scotch-Weld™ metal primer is a one-component, metal and glass primer designed for applications using 3M™ Scotch-Weld™ film and liquid structural adhesives.
  • Improves glass and metal adhesion
  • Resistance to environmental exposure for epoxy and urethane structural adhesives
  • Ensures complete wetting of film adhesive to adhered surfaces
  • Easy application and quick drying
Formulated to improve metal and glass adhesion, this film and liquid adhesive primer simplifies production scheduling by protecting the cleaned surfaces until the bonding operations can be completed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty