Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium glutamate ≥98%

Supplier: Pfaltz & Bauer
Warning
Synonyms: MSGMonosodium glutamateSodium (S)-2-amino-4-carboxybutanoateL-Glutamic acid monosodium salt

S05495-500G
101183-584EA 266.12 USD InStock
101183-584
Sodium glutamate ≥98%
Sodium glutamate


Formula: C₅H₈NNaO₄
MW: 169.112 g/mol
Melting Pt: 232 °C
MDL Number: MFCD00013074
CAS Number: 142-47-2
EINECS: 205-538-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty