Would you like to log in again?
Centrifuge Tubes, Conical Bottom, with Screw Cap, EPPENDORF
Supplier: Eppendorf
0030122160 0030122186
76200-146EA 224.21 USD InStock
76200-146 76200-144
Centrifuge Tubes, Conical Bottom, with Screw Cap, EPPENDORF
Tubes Centrifuge Tubes
These centrifuge tubes are made of Polypropylene, with HDPE scew cap.
  • High quality material and precision molding ensures a high tightness and minimizes sample loss
  • Supports a User-friendly, ergonomic handling (easy opening/closing) and one-handed operation
  • Reliable and safe labeling
  • Each individual lot is checked and certified for quality and purity
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free
Safe design and easy handling ensure best sample processing.
Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity, high g-Safe® centrifugation stability, and optimal sample and pellet visibility.
Ideal for convenient and safe cell culture applications, sample preparation in microbiology and molecular biology labs, mastermixes and buffers preparation, storage of samples and solution with larger volumes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty